Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CCL3/MIP-1 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6625
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Rat samples. It is validated for Western Blot.Specifications
| CCL3/MIP-1 alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P50229 | |
| CCL3 | |
| E. coli-derived recombinant rat CCL3/MIP-1 alpha Ala24-Ala92 Accession # P50229 | |
| 100 μg | |
| Primary | |
| Detects rat CCL3/MIP-1 alpha in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse CCL3 and recombinant cotton rat CCL3 is observed and no cross-reactivity with recombinant human CCL3, recombinant canine CCL3, or recombinant rat CCL4/MIP-1b is observed. | |
| Rat | |
| Purified |
| Western Blot | |
| 755012 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6348 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction