Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CCL3/MIP-1 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB66251SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB66251SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB66251SP Supplier R&D Systems Supplier No. MAB66251SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen CCL3/MIP-1 alpha
Applications Immunohistochemistry
Classification Monoclonal
Clone 769303
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P50229
Gene Alias C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Gene Symbols CCL3
Host Species Mouse
Immunogen E. coli-derived recombinant rat CCL3/MIP-1 alpha Ala24-Ala92 Accession # P50229
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Test Specificity Detects rat CCL3/MIP-1 alpha in direct ELISAs. In direct ELISAs, less than 50% cross-reactivity with recombinant mouse and cotton rat CCL3/MIP-1 alpha is observed, and no cross-reactivity with recombinant human, canine, and HHV CCL3/MIP-1 alpha, recombinant rat CCL4/MIP-1 beta, or recombinant mouse CCL5/RANTES is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.