Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CCR2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$210.00
Specifications
Antigen | CCR2 |
---|---|
Clone | 890231 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB8368R025
![]() |
R&D Systems
FAB8368R025UG |
25 μg |
Each of 1 for $210.00
|
|
|||||
Description
CCR2 Monoclonal specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry.Specifications
CCR2 | |
0.2 mg/mL | |
Monoclonal | |
Mouse | |
Rat | |
O55193 | |
729230 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant |
890231 | |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
CCR2 | |
Primary | |
Stains rat CCR2 transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title