Learn More
Rat CD161 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD161 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Oct-78 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR |
| Host Species | Mouse |
| Immunogen | Purified splenic NK cells from the LEW rat strain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.