Learn More
Rat CD161 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB9766G100UG
Catalog No. FAB9766G100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD161 Monoclonal specifically detects CD161 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD161
Monoclonal
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects rat CD161 in ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
Oct-78
Flow Cytometry 0.25-1 ug/10^6 cells
CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR
Purified splenic NK cells from the LEW rat strain
100 μg
3820
Rat
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.