Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CD19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$227.50 - $514.50
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 771404R |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7489RSP
![]() |
R&D Systems
MAB7489RSP |
25 μg |
Each for $227.50
|
|
|||||
MAB7489R100
![]() |
R&D Systems
MAB7489R100 |
100 μg |
Each for $514.50
|
|
|||||
Description
CD19 Monoclonal specifically detects CD19 in Rat samples. It is validated for Immunohistochemistry.Specifications
| CD19 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
| Chinese hamster ovary cell line CHO-derived recombinant rat CD19 Met1-Gly287 Accession # NP_001013255 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat CD19 in direct ELISAs. |
| 771404R | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| NP_001013255 | |
| 930.0 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title