Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CD25/IL-2R alpha Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB51561T100UG
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Rat samples. It is validated for Flow Cytometry.Specifications
CD25/IL-2R alpha | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P26897 | |
IL2RA | |
Chinese hamster ovary cell line CHO-derived recombinant rat CD25/IL-2 R alpha Glu22-Gln235 Accession # P26897 | |
RUO | |
3559 | |
Rat | |
IgG1 |
Flow Cytometry | |
745520 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
Mouse | |
100 μg | |
Primary | |
Detects rat CD25/IL-2 R alpha in ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) CD25/IL-2 R alpha, rmIL-2 R beta, rmCommon gamma chain, rmIL-15 R alpha, or recombinant human CD25/IL-2 R alpha is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction