Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CD4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6439SP
This item is not returnable.
View return policy
Description
CD4 Polyclonal specifically detects CD4 in Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Sheep | |
Affinity Purified | |
RUO | |
920 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P05540 | |
CD4 | |
Mouse myeloma cell line NS0-derived recombinant rat CD4 Lys28-Thr394 Accession # P05540 | |
25 μg | |
Primary | |
Detects rat CD4 in direct ELISAs and Western blots. In direct ELISAs, approximately 3% cross-reacitvity with recombinant feline CD4 is observed, and less than 1% cross-reactivity with recombinant human CD4 and recombinant canine CD4 is observed. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction