Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CNTF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB557
This item is not returnable.
View return policy
Description
CNTF Monoclonal specifically detects CNTF in Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CNTF | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ciliary neurotrophic factor, HCNTF | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
1270 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Neutralization, ELISA | |
34738 | |
Neutralization 1-3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P20294.1 | |
CNTF | |
E. coli-derived recombinant rat CNTF Ala2-Met200 Accession # P20294.1 | |
500 μg | |
Primary | |
Detects rat CNTF in ELISAs and Western blots. In sandwich immunoassays, less than 3% cross-reactivity with recombinant human CNTF is observed. | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction