Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CNTF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB557SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB557SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB557SP Supplier R&D Systems Supplier No. MAB557SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CNTF Monoclonal specifically detects CNTF in Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CNTF
Applications Neutralization, ELISA
Classification Monoclonal
Clone 34738
Conjugate Unconjugated
Dilution Neutralization 1-3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P20294.1
Gene Alias ciliary neurotrophic factor, HCNTF
Gene Symbols CNTF
Host Species Mouse
Immunogen E. coli-derived recombinant rat CNTF Ala2-Met200 Accession # P20294.1
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1270
Test Specificity Detects rat CNTF in ELISAs and Western blots. In sandwich immunoassays, less than 3% cross-reactivity with recombinant human CNTF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.