Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CNTFR alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF559NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF559NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF559NA Supplier R&D Systems Supplier No. AF559NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CNTFR alpha Polyclonal specifically detects CNTFR alpha in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen CNTFR alpha
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 2-6 ug/mL
Gene Accession No. Q08406
Gene Symbols Cntfr
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat CNTF R alpha Ala19-Pro346 Accession # Q08406
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1271
Test Specificity Detects human CNTF R alpha in direct ELISAs and Western blots. In direct ELISAs, greater than 50% cross-reactivity with recombinant human CNTF sR alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.