Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat Complement C4a ELISA Kit (Colorimetric)

Click to view available options
Quantity:
1 Kit
Description
The Rat Complement C4a ELISA Kit (Colorimetric) measures rat Complement C4a in serum, plasma and other biological fluids.
Specifications
Specifications
Assay Range | 0.31 - 20 ng/mL |
Assay Sensitivity | 0.19 ng/mL |
Cross Reactivity | This kit recognizes Rat C4a in samples. No significant cross-reactivity or interference between Rat C4a and analogues was observed. |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Biological Fluids, Plasma, Serum |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 720 |
Gene Symbol | C4A |
Host Species | Rat |
Quantity | 1 Kit |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction