Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Contactin-1 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP275803 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275803 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275803 Supplier Novus Biologicals™ Supplier No. NBP275803
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Rat Contactin-1 ELISA Kit (Colorimetric) measures rat Contactin-1 in serum, plasma and other biological fluids.

Specifications

Assay Range 0.16 - 10 ng/mL
Assay Sensitivity 0.10 ng/mL
Cross Reactivity This kit recognizes Rat CNTN1 in samples. No significant cross-reactivity or interference between Rat CNTN1 and analogues was observed.
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 1272
Gene Symbol CNTN1
Host Species Rat
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Contactin-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.