Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CX3CL1/Fractalkine Chemokine Domain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB537
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB537 500 μg
MAB537SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB537 Supplier R&D Systems Supplier No. MAB537
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Rat samples. It is validated for Western Blot.

Specifications

Antigen CX3CL1/Fractalkine
Applications Western Blot
Classification Monoclonal
Clone 96834
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O55145
Gene Alias ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1
Gene Symbols CX3CL1
Host Species Mouse
Immunogen E. coli-derived recombinant rat CX3CL1/Fractalkine Chemokine Domain Gln25-Gly100 Accession # O55145
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6376
Test Specificity Detects rat CX3CL1/Fractalkine Chemokine in direct ELISAs and Western blots. In ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) CX3CL1 is observed, 10% cross-reactivity with rmCCL11 is observed, and no cross-reactivity with recombinant human (rh) CX3CL1 or rhCCL11 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.