Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CX3CL1/Fractalkine Chemokine Domain Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF537
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF537 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF537 Supplier R&D Systems Supplier No. BAF537
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CX3CL1/Fractalkine Polyclonal specifically detects CX3CL1/Fractalkine in Rat samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen CX3CL1/Fractalkine
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. O55145
Gene Alias ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1
Gene Symbols CX3CL1
Host Species Goat
Immunogen E. coli-derived recombinant rat CX3CL1/Fractalkine Chemokine Domain Gln25-Gly100 Accession # O55145
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6376
Test Specificity Detects rat CX3CL1/Fractalkine Chemokine Domain in ELISAs and Western blots. In sandwich ELISAs, approximately 30% cross-reactivity with recombinant human (rh) Fractalkine (362-CX) is observed and approximately 5% cross-reactivity with recombinant mouse (rm) Fractalkine (571-MF) and rmFractalkine (472-FF) and less than 2% cross-reactivity with rmFractalkine (458-MF) and rhFractalkine (365-FR) is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.