Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB515100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB515100 100 μg
MAB515SP 25 μg
MAB515500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB515100 Supplier R&D Systems Supplier No. MAB515100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CXCL1/GRO alpha/KC/CINC-1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 251021
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 3-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P14095
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Gene Symbols Cxcl1
Host Species Mouse
Immunogen E. coli-derived recombinant rat CXCL1/GRO alpha/KC/CINC-1 Ala25-Lys96 Accession # P14095
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Test Specificity Detects rat CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human (rh) GRO alpha , rhGRO beta , rhGRO gamma , and recombinant mouse KC is observed. No cross-reactivity with recombinant rat (rr) CINC2a, rrCXCL3, rhCXCL5, 6, 7, 8, 9, 10, 11, 12 or 13, rmCXCL2, 6, 9, 10, 12, 13, or recombinant porcine CXCL8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.