Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF515NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF515NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF515NA Supplier R&D Systems Supplier No. AF515NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

CXCL1/GRO alpha/KC/CINC-1 Polyclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen CXCL1/GRO alpha/KC/CINC-1
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.15-0.75 ug/mL
Gene Accession No. P14095
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Gene Symbols Cxcl1
Host Species Goat
Immunogen E. coli-derived recombinant rat CXCL1/GRO alpha/KC/CINC-1, Ala25-Lys96, Accession # P14095
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Test Specificity Detects rat CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse (rm) KC is observed and less than 10% cross-reactivity with recombinant human (rh) GRO beta , rhGRO gamma , r
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.