Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat CXCL16 ELISA Kit (Colorimetric)

Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Assay Range | 0.16 to 10 ng/mL |
Assay Sensitivity | 0.09 ng/mL |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum |
Specificity | This kit recognizes Rat CxCL16 in samples. No Significant cross-reactivity or interference between Rat CxCL16 and analogues was observed. |
Target Species | Rat |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 58191 |
Gene Symbol | CXCL16 |
Quantity | 1 Kit |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction