Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CXCL2/GRO beta/MIP-2/CINC-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB525SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB525SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB525SP Supplier R&D Systems Supplier No. MAB525SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CXCL2/GRO beta/MIP-2/CINC-3 Monoclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Rat samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL2/GRO beta/MIP-2/CINC-3
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 123802
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.4-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P30348
Gene Alias chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2
Gene Symbols CXCL2
Host Species Mouse
Immunogen E. coli-derived recombinant rat CXCL2/GRO beta/MIP-2/CINC-3 Ser32-Asn100 Accession # P30348
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2920
Test Specificity Detects rat CXCL2/GRO beta/MIP-2/CINC-3 in ELISAs and Western blots. In sandwich ELISAs, less than 0.5% cross-reactivity with recombinant rat (rr) CXCL3 and less than 0.1% cross-reactivity with rrCXCL1, rrCINC-2 alpha , recombinant mouse (rm) CXCL9 or rmCXCL2 is observed. In Western blots, less than 3% cross-reactivity with recombinant human (rh) CXCL8, recombinant porcine CXCL8, or rrCINC-2 alpha and no cross-reactivity with rrCXCL1, 3, rmCXCL1, 2, 6, 9, 10, 12, rhCXLC1, 2, 3, 5, 6, 7, 9,10, 11, 12 or 13 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.