Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CXCL3/CINC-2 alpha/beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF517SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF517SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF517SP Supplier R&D Systems Supplier No. AF517SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CXCL3/GRO gamma/CINC-2/DCIP-1 Polyclonal specifically detects CXCL3/GRO gamma/CINC-2/DCIP-1 in Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CXCL3/GRO gamma/CINC-2/DCIP-1
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.4-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_612531
Gene Alias chemokine (C-X-C motif) ligand 3, CINC-2, CINC-2b, C-X-C motif chemokine 3, Dcip1, Gm1960, GRO gamma, GRO3, GRO3 oncogene, GROg, GRO-gamma, GRO-gamma(1-73), Growth-regulated protein gamma, Macrophage inflammatory protein 2-beta, melanoma growth stimulatory activity gamma, MGSA gamma, MIP-2b, MIP2B, MIP2-beta, SCYB3
Gene Symbols Cxcl3
Host Species Goat
Immunogen E. coli-derived recombinant rat CXCL3/CINC-2 alpha/beta Arg33-Leu100 Accession # NP_612531
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2921
Test Specificity Detects rat CXCL3/CINC-2 alpha/beta in direct ELISAs and Western blots. In direct ELISAs, almost 100% cross-reactivity with recombinant rat (rr) CINC-2 alpha is observed, approximately 50% cross-reactivity with recombinant mouse (rm) MIP-2 is observed, 35% cross-reactivity with rmKC is observed, 15% cross-reactivity with rrCINC-1 is observed, and 10% cross-reactivity with recombinant human (rh) IL-8, rhGRO alpha , rhGRO beta , and rhGRO gamma is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.