Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CXCL7/Thymus Chemokine-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1116
This item is not returnable.
View return policy
Description
CXCL7/Thymus Chemokine-1 Monoclonal specifically detects CXCL7/Thymus Chemokine-1 in Rat samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CXCL7/Thymus Chemokine-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 4-20 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
NP_714943 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5473 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Neutralization, ELISA | |
186701 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Ppbp | |
E. coli-derived recombinant rat CXCL7/Thymus Chemokine-1 Ile46-Ile107 Accession # NP_714943 | |
500 μg | |
Primary | |
Detects rat CXCL7/Thymus Chemokine-1 in ELISAs and Western blots. In sandwich ELISAs, no cross-reactivity or interference with recombinant human CXCL5, recombinant mouse CXCL7/Thymus Chemokine-1, recombinant rat (rr) CXCL1/CINC1, rrCXCL2/CINC3, rrCXCL3/CINC-2 alpha, rrCXCL3/CINC-2 beta, or rrCXCL5 is observed. | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction