Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CXCL7/Thymus Chemokine-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1116
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1116 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1116 Supplier R&D Systems Supplier No. AF1116
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CXCL7/Thymus Chemokine-1 Polyclonal specifically detects CXCL7/Thymus Chemokine-1 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen CXCL7/Thymus Chemokine-1
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.8-4.0 ug/mL
Gene Accession No. NP_714943
Gene Symbols Ppbp
Host Species Goat
Immunogen E. coli-derived recombinant rat CXCL7/Thymus Chemokine-1 Ile46-Ile107 Accession # NP_714943
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5473
Test Specificity Detects CXCL7/Thymus Chemokine-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse CXCL7 is observed and less than 1% cross-reactivity with other chemokines tested.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.