Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CXCR6 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8196N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8196N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8196N100 Supplier R&D Systems Supplier No. FAB8196N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCR6 Monoclonal specifically detects CXCR6 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR6
Applications Flow Cytometry
Classification Monoclonal
Clone 879112
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186
Gene Symbols CXCR6
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with rat CXCR6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10663
Test Specificity Detects rat CXCR6 in ELISA. Stains HEK293 cells transfected with rat CXCR6 by Flow Cytometry, but does not stain irrelevant transfectants.
Target Species Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.