Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat D-Dimer ELISA Kit (Colorimetric)
SDP

Catalog No. NB398076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398076 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398076 Supplier Novus Biologicals™ Supplier No. NBP308101
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 78.13 to 5000 ng/mL
Assay Sensitivity 46.88 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma
Target Species Rat
For Use With (Application) ELISA
Quantity 1 Kit
Research Discipline Extracellular Matrix
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym D Dimer
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.