Learn More
Specifications
Specifications
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 37.5 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum, Plasma, Other Biological Fluids |
| Specificity | This kit recognizes Rat Defensin 5 in samples. No significant cross reactivity or interference between Rat Defensin 5 and analogues was observed. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Rat |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
