Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Dkk-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4010
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4010 100 μg
AF4010SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4010 Supplier R&D Systems Supplier No. AF4010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Dkk-1 Polyclonal specifically detects Dkk-1 in Rat samples. It is validated for Western Blot.

Specifications

Antigen Dkk-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_001099820
Gene Alias dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like
Gene Symbols DKK1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Dkk-1 (R&D Systems, Catalog # 4010-DK) Ser23-His270 Accession # NP_001099820
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22943
Test Specificity Detects rat Dkk-1 in direct ELISAs and Western blots. In these formats, approximately 40% cross-reactivity with recombinant mouse Dkk-1 is observed and approximately 20% cross-reactivity with recombinant human Dkk-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.