Learn More
Description
- The Rat Dkk-1 ELISA Kit (Colorimetric) measures rat Dkk-1 in culture supernatants, plasma, and serum.
- Assay Type: Sandwich ELISA
- Standard Curve Range: 39.1 - 1250 pg/mL
- Sensitivity: 26.1 pg/mL
- Inter-Assay: Mean 9.9%
- Intra-Assay: Mean 8.0%
- Spike Recovery:
- F12K + 10% FBS
- Low - 88%
- Medium - 96%
- High - 109%
- Serum
- Low - 77
- Medium - 101%
- High - 116%
- EDTA Plasma
- Low - 92%
- Medium - 98%
- High - 98%
- F12K + 10% FBS
- Sample Volume: 100 μL
- Components:
- Antibody
- Assay buffer 36
- Conjugate
- Standard
- Microtiter plate
- Stop solution 2
- TMB Substrate
- Wash buffer concentrate
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 22943 |
| Gene Symbol | DKK1 |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
