Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat DLL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3970
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3970 100 μg
MAB3970SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3970 Supplier R&D Systems Supplier No. MAB3970
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DLL1 Monoclonal specifically detects DLL1 in Rat samples. It is validated for Western Blot.

Specifications

Antigen DLL1
Applications Western Blot
Classification Monoclonal
Clone 409005
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P97677
Gene Alias Delta, delta (Drosophila)-like 1, Delta 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1
Gene Symbols DLL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat DLL1 Gln18-Trp537 Accession # P97677
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28514
Test Specificity Detects rat DLL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant rat (rr) Jagged-1, recombinant mouse (rm) Jagged-2, rmNotch-3, rrNotch-2, and rrNotch-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.