Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat E-Selectin/CD62E Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF977SP
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Polyclonal specifically detects E-Selectin/CD62E in Rat samples. It is validated for Western Blot, Immunohistochemistry, Adhesion Blockade.Specifications
E-Selectin/CD62E | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
Goat | |
Affinity Purified | |
RUO | |
6401 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Adhesion Blockade | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Adhesion Blockade | |
P98105 | |
SELE | |
Mouse myeloma cell line NS0-derived recombinant rat E-Selectin/CD62E Trp22-Pro494 Accession # P98105 | |
25 μg | |
Primary | |
Detects E-Selectin/CD62E in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse (rm) E-Selectin is observed, approximately 5% cross-reactivity with recombinant human (rh) E-Selectin is observed and less than 1% cross-reactivity with rmL-Selectin and rmP-Selectin is observed. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction