Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat E-Selectin ELISA Kit

Catalog No. p-7219880
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests

ELISA

Rat E-Selectin quantitates rat E-Selectin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat E-Selectin.

Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CD62 antigen-like family member E; CD62E; e selectin; Elam; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule 1; ESEL; E-selectin; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2; Sele; selectin E; selectin E (endothelial adhesion molecule 1); selectin, endothelial cell
Assay Range 0.13 to 20 ng/mL
Assay Sensitivity 0.13 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 25544
Gene Symbol SELE
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target E-selectin (CD62E)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.