Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Epidemic Hemorrhagic Fever Virus Antibody (IgG) ELISA Kit (Colorimetric)
SDP

Catalog No. NB599881KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB599881KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB599881KT Supplier Novus Biologicals™ Supplier No. NBP2599881KIT
Add to Cart
Add to Cart

Rat Epidemic Hemorrhagic Fever Virus Antibody (IgG) ELISA Kit (Colorimetric) employs the qualitative enzyme immunoassay technique for the detection of rat epidemic hemorrhagic fever antibody(IgG)

The Rat Epidemic Hemorrhagic Fever Virus Antibody (IgG) ELISA Kit (Colorimetric) measures rat Epidemic Hemorrhagic Fever Virus Antibody (IgG) in serum.

Specifications

Assay Range Qualitative
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
Specificity For detection of rat epidemic hemorrhagic fever antibody(IgG)
Target Species Rat
For Use With (Application) ELISA
Kit Contents Adhesive Strip (For 96 wells), HRP-conjugate, Coated assay plate, Substrate B, Substrate A, Wash Buffer (30 x concentrate), Stop Solution, Sample Diluent, Instruction manual, Positive Control, Negative Control
Quantity 1 Kit
Research Discipline Cell Biology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Epidemic Hemorrhagic Fever Virus Antibody (IgG)
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.