Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Fas Ligand/TNFSF6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1889
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1889 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1889 Supplier R&D Systems Supplier No. BAF1889
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Fas Ligand/TNFSF6 Polyclonal specifically detects Fas Ligand/TNFSF6 in Rat samples. It is validated for Western Blot.

Specifications

Antigen Fas Ligand/TNFSF6
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P36940
Gene Alias apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Gene Symbols FASLG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Fas Ligand/TNFSF6 Leu104-Leu278 Accession # P36940
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 356
Test Specificity Detects rat Fas Ligand/TNFSF6 in Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse Fas Ligand and 10% cross-reactivity with recombinant human Fas Ligand is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.