Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Fetuin A/AHSG Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7510SP
This item is not returnable.
View return policy
Description
Fetuin A/AHSG Monoclonal specifically detects Fetuin A/AHSG in Rat samples. It is validated for Western Blot.Specifications
Fetuin A/AHSG | |
Monoclonal | |
LYOPH | |
Western Blot 0.2 ug/mL | |
P24090 | |
AHSG | |
Mouse myeloma cell line NS0-derived recombinant rat Fetuin A/AHSG Ala19-I352 Accession # P24090 | |
25 μg | |
Primary | |
Detects rat Fetuin A/AHSG in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Fetuin A or recombinant mouse Fetuin A is observed. | |
Rat | |
Purified |
Western Blot | |
774761 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
A2HS, AHS, AHSG, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
197 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction