Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat GFR alpha-1/GDNF R alpha-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB560SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB560SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB560SP Supplier R&D Systems Supplier No. MAB560SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

GFR alpha-1/GDNF R alpha-1 Monoclonal specifically detects GFR alpha-1/GDNF R alpha-1 in Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GFR alpha-1/GDNF R alpha-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 81401
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q62997
Gene Alias DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF R alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR alpha1, GFRa-1, GFRA1, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546
Gene Symbols GFRA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat GFR alpha-1/GDNF R alpha-1 Asp25-Leu445 Accession # Q62997
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2674
Test Specificity Detects rat GFR alpha-1/GDNF R alpha-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Ret, recombinant human (rh) GFR alpha-2, rhGFR alpha-3 or rmGFR alpha-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.