Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Glut4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1262
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1262 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1262 Supplier R&D Systems Supplier No. MAB1262
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 13 publications

Glut4 Monoclonal specifically detects Glut4 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification, Immunocytochemistry.

Specifications

Antigen Glut4
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification, Immunocytochemistry
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 5-25 μg/mL, Immunoprecipitation, Immunoaffinity Purification, Immunocytochemistry 8-25 μg/mL
Formulation Supplied as a 0.2 um filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, SLC2A4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4
Gene Symbols SLC2A4
Host Species Mouse
Immunogen Partially purified vesicles containing Glut4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6517
Test Specificity Recognizes an epitope in the cytoplasmic portion of rat Glut4 and has been shown to bind only to the muscle-adipose isoform of the glucose transporter, which is antigenically unique from the other transport proteins (1-3). Recognizes Glut4 in human, monkey, rat, mouse, and rabbit systems. It does not recognize this protein in canine systems. Its ability to bind to glucose transporter from other species has not been tested.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.