Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Glycine R Pan Specific Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems PPS009
This item is not returnable.
View return policy
Description
Glycine R Polyclonal specifically detects Glycine R in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Glycine R | |
Polyclonal | |
Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 dilution | |
Peptide from the N-terminus region of the alpha 1 subunit of the rat Glycine Receptor | |
200 μg | |
Primary | |
This antibody should be reconstituted in 100 μL phosphate buffered saline (137 mM NaCl, 7.5 mM Na2HPO4, 2.7 mM KCl, 1.5 mM KH2PO4, pH 7.4) before use. | |
The lyophilized product is stable at ≤−20degrees C for at least 1 year. After reconstitution the antibody should be aliquoted and stored at ≤ −20°C. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Specific for the ∼48 kDa alpha 1 and alpha 2 subunits of the Glycine Receptor in Western blots of rat spinal cord, brain stem and cell extracts. Immunolabeling is blocked by preadsorption of antibody with the peptide immunogen. This antibody does not recognize other Glycine Receptor subunits. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction