Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat GM-CSF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB518SP
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
GM-CSF | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
1437 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Neutralization, ELISA | |
83329 | |
Neutralization 0.2-0.8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P48750 | |
CSF2 | |
E. coli-derived recombinant rat GM-CSF Ala18-Lys144 Accession # P48750 | |
25 μg | |
Primary | |
Detects rat GM-CSF in ELISAs. | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction