Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat gp130 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5029G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5029G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5029G100 Supplier R&D Systems Supplier No. FAB5029G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

gp130/CD130 Monoclonal specifically detects gp130/CD130 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen gp130/CD130
Applications Flow Cytometry
Classification Monoclonal
Clone 745314
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P40190
Gene Alias CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
Gene Symbols IL6ST
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat gp130 Gln23-Glu618 (predicted) Accession # P40190
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3572
Test Specificity Detects rat gp130 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human or recombinant mouse gp130 is observed.
Target Species Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.