Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat gp130 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5029N100UG
This item is not returnable.
View return policy
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Rat samples. It is validated for Flow Cytometry.Specifications
gp130/CD130 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P40190 | |
IL6ST | |
Mouse myeloma cell line NS0-derived recombinant rat gp130 Gln23-Glu618 (predicted) Accession # P40190 | |
RUO | |
3572 | |
Rat | |
IgG1 |
Flow Cytometry | |
745314 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
Mouse | |
100 μg | |
Primary | |
Detects rat gp130 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human or recombinant mouse gp130 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction