Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat gp130 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5029
This item is not returnable.
View return policy
Description
gp130/CD130 Polyclonal specifically detects gp130/CD130 in Rat samples. It is validated for Western Blot.Specifications
gp130/CD130 | |
Polyclonal | |
Unconjugated | |
EDM10349 | |
IL6ST | |
Mouse myeloma cell line NS0-derived recombinant rat gp130 Gln23-Glu618, predicted Accession # EDM10349 | |
100 μg | |
Primary | |
Detects rat gp130 in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse gp130 is observed, and less than 6% cross-reactivity with recombinant human gp130 is observed. | |
Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL | |
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
Sheep | |
Affinity Purified | |
RUO | |
3572 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction