Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Growth Hormone Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB15661
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB15661 500 μg
MAB15661SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB15661 Supplier R&D Systems Supplier No. MAB15661
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Growth Hormone Monoclonal specifically detects Growth Hormone in Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Growth Hormone
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 222517
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01244
Gene Alias GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin
Gene Symbols GH1
Host Species Mouse
Immunogen E. coli-derived recombinant rat Growth Hormone Phe27-Phe216 Accession # P01244
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2688
Test Specificity Detects rat Growth Hormone in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Growth Hormone is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.