Learn More
Description
Specifications
Specifications
| Assay Range | 125.00 to 8000 pg/mL |
| Assay Sensitivity | 75.00 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat HSP-90 in samples. No significant cross-reactivity or interference between Rat HSP-90 and analogues was observed. |
| Gene ID (Entrez) | 3320 |
| Gene Symbol | HSP90AA1 |
| Host Species | Rat |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
