Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IFN-beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. 334001
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
334001 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 334001 Supplier R&D Systems Supplier No. 334001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFN-beta Polyclonal specifically detects IFN-beta in Rat samples. It is validated for Neutralization.

Specifications

Antigen IFN-beta
Applications Neutralization
Classification Polyclonal
Conjugate Unconjugated
Dilution Neutralization
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Gene Symbols IFNB1
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant rat IFN-beta
Purification Method Affinity Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Test Specificity Neutralizes both natural and recombinant rat interferon beta. Cross-reactivity with human interferon beta has not been tested.
Reconstitution Dissolve contents of the vial by injection of 0.5 ml sterile distilled water.
Target Species Rat
Content And Storage Upon receipt, this product is stable at 4degreesC until the expiration date listed above. After reconstitution, the contents can be safely stored at 4degreesC for one month or at -20degreesC for one year for retention of full activity.When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Add 0.02% sodium azide to prevent bacterial growth.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.