Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat IFN-gamma ELISA Kit (Colorimetric)
SDP

Catalog No. nbp192680
Click to view available options
Quantity:
96 Tests

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Rat IFN-gamma ELISA Kit (Colorimetric) measures rat IFN-gamma in cell culture supernatant, serum.

Specifications

Assay Range 31.3 - 2,000pg/mL
Assay Sensitivity 9.9pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Serum
Specificity The interference of circulating factors of the immune systemwas evaluated by spiking these proteins at physiologically relevant concentrations into a rat IFN gammapositive serum. There was no crossreactivity detected.
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 3458
Gene Symbol IFNG
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IFN-gamma, Biotin-Conjugate anti-rat IFN-gamma monoclonal antibody, Streptavidin-HRP, Rat IFN-gamma Standard lyophilized, 4ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Target IFN-gamma
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.