Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat IGFBP-3 ELISA Kit (Colorimetric)
SDP

Catalog No. NB013312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013312 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013312 Supplier Novus Biologicals™ Supplier No. NBP280241

This kit recognizes Rat IGFBP-3 in samples.

No significant cross-reactivity or interference between Rat IGFBP-3 and analogues was observed.

Specifications

Assay Range 78.125 to 5000 pg/mL
Assay Sensitivity 46.875 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 3486
Gene Symbol IGFBP3
Host Species Rat
Quantity 1 Kit
Research Discipline Apoptosis, Biologically Active Proteins, Prostate Cancer
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target IGFBP-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.