Learn More
Rat IgG1 Kappa Light Chain, Clone: RG1, APC, Isotype control, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | IgG1 Kappa Light Chain |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RG1 |
| Conjugate | APC |
| Dilution | Flow Cytometry 1:10 - 1:1000 |
| Formulation | Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide |
| Host Species | Rat |
| Quantity | 0.05 mg |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.