Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG1 Kappa Light Chain Isotype Control (KLH/G1-2-2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP151817W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151817W 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151817W Supplier Novus Biologicals Supplier No. NBP151817AF405
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG1 Kappa Light Chain Monoclonal specifically detects IgG1 Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, ELISA, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG1 Kappa Light Chain
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone KLH/G1-2-2
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Formulation 50 mM Sodium Borate. with 0.05% Sodium Azide
Host Species Rat
Immunogen Keyhole limpet hemocyanin (KLH)
Quantity 500 Tests
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Test Specificity Immunoglobulin reacts with Keyhole limpet hemocyanin (KLH).
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.