Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG1 Kappa Light Chain Isotype Control (RG1), PerCP/Cy5.5, Novus Biologicals™
SDP

Catalog No. NBP142841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142841 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142841 Supplier Novus Biologicals Supplier No. NBP142841
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG1 Kappa Light Chain Monoclonal specifically detects IgG1 Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG1 Kappa Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone RG1
Conjugate PerCP-Cyanine5.5
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Host Species Rat
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Rat
Content And Storage Store at 4C in the dark.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.