Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2a Isotype Control (KLH/G2a-1-1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP151104IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151104IR 250 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151104IR Supplier Novus Biologicals Supplier No. NBP151104IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG2a Monoclonal specifically detects IgG2a in Rat samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen IgG2a
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone KLH/G2a-1-1
Conjugate DyLight 755
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000
Formulation 50 mM Sodium Borate. with 0.05% Sodium Azide
Host Species Rat
Immunogen Keyhole limpet hemocyanin (KLH)
Quantity 250 Tests
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Test Specificity Immunoglobulin reacts with Keyhole limpet hemocyanin (KLH).
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.