Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2b Isotype Control, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. DDXCR03A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDXCR03A488 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDXCR03A488 Supplier Novus Biologicals Supplier No. DDXCR03A488
Add to Cart
Add to Cart

Rat Polyclonal Antibody

IgG2b Polyclonal specifically detects IgG2b in Rat samples. It is validated for Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence.

Specifications

Antigen IgG2b
Applications Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Dilution Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500
Formulation PBS, 50% glycerol. with No Preservative
Host Species Rat
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.